Gene Transcripts

Gene Symbol
Lrp2
Entrez Gene ID
29216
Gene Name
low density lipoprotein receptor-related protein 2
Gene Aliases
Chromosome Location
Chr. 3: 55665153 - 55822484 on Build Rnor_6.0
UniGene ID
Rn.26430
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030827.1 NP_110454.1 50 9894
GenBank mRNA L34049.1 - 50 9894

View Full Product Details