Gene Transcripts

Gene Symbol
Opn1sw
Entrez Gene ID
81644
Gene Name
opsin 1 (cone pigments), short-wave-sensitive
Gene Aliases
Chromosome Location
Chr. 4: 56653840 - 56656980 on Build Rnor_6.0
UniGene ID
Rn.10549
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031015.1 NP_112277.1 1 288
GenBank mRNA AF051163.1 - 1 240
U63972.1 - 1 288

View Full Product Details