Gene Transcripts

Gene Symbol
Akr1a1
Entrez Gene ID
78959
Gene Name
aldo-keto reductase family 1, member A1 (aldehyde reductase)
Gene Aliases
Akr1a4
Chromosome Location
Chr. 5: 135482068 - 135498693 on Build Rnor_6.0
UniGene ID
Rn.835
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031000.3 NP_112262.1 7, 8 886
GenBank mRNA BC059133.1 - 7, 8 1165
D10854.1 - 7, 8 886
FQ212523.1 - 7, 8 1129
FQ212586.1 - 7, 8 1126
FQ212779.1 - 7, 8 1161
FQ212919.1 - 6, 7 836
FQ213467.1 - 7, 8 1161
FQ213483.1 - 7, 8 1157
FQ213491.1 - 7, 8 1159
FQ213579.1 - 7, 8 1129
FQ213776.1 - 7, 8 1128
FQ214173.1 - 7, 8 1129
FQ214441.1 - 7, 8 905
FQ214642.1 - 1142
FQ214700.1 - 7, 8 1102
FQ215620.1 - 7, 8 953
FQ215772.1 - 7, 8 1129
FQ218383.1 - 7, 8 1129
FQ218721.1 - 7, 8 1128
FQ218729.1 - 7, 8 1128
FQ220055.1 - 5, 6 729
FQ228132.1 - 7, 8 1161
FQ228842.1 - 7, 8 1157
FQ229180.1 - 6, 7 800
FQ233156.1 - 7, 8 986

View Full Product Details