Gene Transcripts

Gene Symbol
Chrm2
Entrez Gene ID
81645
Gene Name
cholinergic receptor, muscarinic 2
Gene Aliases
Acm2
Chromosome Location
Chr. 4: 64089028 - 64091090 on Build Rnor_6.0
UniGene ID
Rn.10752
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031016.1 NP_112278.1 1 1171
GenBank mRNA AB017655.1 - 1 720
J03025.1 - 1 1171

View Full Product Details