Gene Transcripts

Gene Symbol
ONECUT2
Entrez Gene ID
9480
Gene Name
one cut homeobox 2
Gene Aliases
OC-2, OC2
Chromosome Location
Chr. 18: 57435420 - 57491298 on Build GRCh38
UniGene ID
Hs.194725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004852.2 NP_004843.2 1 1209
XM_017026096.1 XP_016881585.1 1 1474
GenBank mRNA BC108925.1 - 2 1034
BC108926.1 - 2 1034
Y18198.1 - 1 1208

View Full Product Details