Gene Transcripts
- Gene Symbol
- TAX1BP1
- Entrez Gene ID
- 8887
- Gene Name
- Tax1 binding protein 1
- Gene Aliases
- CALCOCO3, T6BP, TXBP151
- Chromosome Location
- Chr. 7: 27739373 - 27829767 on Build GRCh38
- UniGene ID
- Hs.34576
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001079864.2 |
NP_001073333.1
|
2, 3 |
352 |
|
NM_001206901.1 |
NP_001193830.1
|
2, 3 |
355 |
|
NM_001206902.1 |
NP_001193831.1
|
2, 3 |
352 |
|
NM_006024.6 |
NP_006015.4
|
2, 3 |
352 |
|
XM_005249900.2 |
XP_005249957.1
|
2, 3 |
352 |
|
XM_017012754.1 |
XP_016868243.1
|
2, 3 |
352 |
|
GenBank mRNA |
AF090891.1 |
- |
2, 3 |
270 |
|
AF268075.1 |
- |
1, 2 |
170 |
|
AK000098.1 |
- |
2, 3 |
265 |
|
AK296720.1 |
- |
2, 3 |
301 |
|
AK299729.1 |
- |
2, 3 |
355 |
|
AK301064.1 |
- |
2, 3 |
291 |
|
AK314292.1 |
- |
2, 3 |
322 |
|
AL136586.1 |
- |
2, 3 |
258 |
|
BC050358.2 |
- |
2, 3 |
237 |
6055189 |
BP280714.1 |
- |
2, 3 |
328 |
|
CR457056.1 |
- |
1, 2 |
170 |
|
CR533556.1 |
- |
1, 2 |
170 |
|
DB455556.1 |
- |
2, 3 |
351 |
|
DC424592.1 |
- |
2, 3 |
349 |
|
U33821.2 |
- |
2, 3 |
253 |
|
View Full Product Details