Gene Transcripts

Gene Symbol
TFDP2
Entrez Gene ID
7029
Gene Name
transcription factor Dp-2
Gene Aliases
DP2
Chromosome Location
Chr. 3: 141944428 - 142149544 on Build GRCh38
UniGene ID
Hs.379018
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178138.1 NP_001171609.1 8 870
NM_001178139.1 NP_001171610.1 7 835
NM_001178140.1 NP_001171611.1 4 331
NM_001178141.1 NP_001171612.1 4 264
NM_001178142.1 NP_001171613.1 2 112
NM_006286.4 NP_006277.1 5 386
XM_005247731.2 XP_005247788.1 7 835
XM_011513101.2 XP_011511403.1 8 931
XM_011513102.2 XP_011511404.1 6 504
XM_011513106.2 XP_011511408.1 8 931
XM_011513107.1 XP_011511409.1 7 835
XM_011513108.2 XP_011511410.1 5 658
XM_011513109.2 XP_011511411.1 3 230
XM_011513111.2 XP_011511413.1 3 230
XM_017007091.1 XP_016862580.1 7 846
XM_017007092.1 XP_016862581.1 10 1089
XM_017007093.1 XP_016862582.1 10 1278
XM_017007094.1 XP_016862583.1 7 864
XM_017007095.1 XP_016862584.1 9 1054
XM_017007096.1 XP_016862585.1 6 761
XM_017007097.1 XP_016862586.1 8 863
XM_017007098.1 XP_016862587.1 8 1008
XM_017007099.1 XP_016862588.1 9 986
XM_017007100.1 XP_016862589.1 7 835
XM_017007101.1 XP_016862590.1 7 816
XM_017007102.1 XP_016862591.1 5 614
XM_017007103.1 XP_016862592.1 7 835
XM_017007104.1 XP_016862593.1 5 658
XM_017007105.1 XP_016862594.1 4 591
GenBank mRNA AK303181.1 - 2 73
AK303634.1 - 4 206
AK314786.1 - 7 692
AL080206.1 - 4 291
AW673504.1 - 4 331
BC021113.1 - 4 290 4653771
DC319314.1 - 5 386
L40386.1 - 4 230
U18422.1 - 5 338

View Full Product Details