Gene Transcripts

Gene Symbol
Tesk1
Entrez Gene ID
29460
Gene Name
testis-specific kinase 1
Gene Aliases
Chromosome Location
Chr. 5: 58937615 - 58943358 on Build Rnor_6.0
UniGene ID
Rn.7006
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031578.1 NP_113766.1 9 2116
GenBank mRNA BC081773.1 - 9 1712
D50864.1 - 9 2116

View Full Product Details