Gene Transcripts

Gene Symbol
Cryba4
Entrez Gene ID
64348
Gene Name
crystallin, beta A4
Gene Aliases
Chromosome Location
Chr. 12: 50407843 - 50414432 on Build Rnor_6.0
UniGene ID
Rn.10802
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031689.1 NP_113877.1 5 360
GenBank mRNA AF013247.1 - 5 360

View Full Product Details