Gene Transcripts

Gene Symbol
Apln
Entrez Gene ID
58812
Gene Name
apelin
Gene Aliases
Apel
Chromosome Location
Chr. X: 134856719 - 134866210 on Build Rnor_6.0
UniGene ID
Rn.22847
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031612.3 NP_113800.1 2 520
GenBank mRNA AB023495.1 - 2 180
AF179679.1 - 2 480
BC080843.1 - 2 506 7099981

View Full Product Details