Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004739.3 | NP_004730.2 | 9 | 1253 | |
XM_017018561.1 | XP_016874050.1 | 9 | 1105 | ||
GenBank mRNA | AB016591.1 | - | 9 | 1186 | |
AF295807.1 | - | 9 | 863 | ||
AK301569.1 | - | 9 | 1088 | ||
BC023656.2 | - | 9 | 1049 | ||
BC053650.1 | - | 9 | 1071 | 5531965 | |
CR749481.1 | - | 7 | 774 |