Gene Transcripts

Gene Symbol
Mlycd
Entrez Gene ID
85239
Gene Name
malonyl-CoA decarboxylase
Gene Aliases
Mcd
Chromosome Location
Chr. 19: 52032950 - 52048773 on Build Rnor_6.0
UniGene ID
Rn.13468
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053477.1 NP_445929.1 2 626
GenBank mRNA AF304865.1 - 2 626
AJ007704.1 - 2 626
BC061845.1 - 2 629 5621015

View Full Product Details