Gene Transcripts

Gene Symbol
Flt4
Entrez Gene ID
114110
Gene Name
fms-related tyrosine kinase 4
Gene Aliases
Vegfr3
Chromosome Location
Chr. 10: 35078782 - 35120296 on Build Rnor_6.0
UniGene ID
Rn.81043
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053652.1 NP_446104.1 2 116
GenBank mRNA AF402785.1 - 2 116
AF402786.1 - 2 116

View Full Product Details