Gene Transcripts
- Gene Symbol
- Ptf1a
- Entrez Gene ID
- 117034
- Gene Name
- pancreas specific transcription factor, 1a
- Gene Aliases
- Chromosome Location
- Chr. 17: 86199623 - 86201477 on Build Rnor_6.0
- UniGene ID
- Rn.10536
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053964.1 |
NP_446416.1
|
1 |
388 |
|
GenBank mRNA |
X98170.1 |
- |
1 |
380 |
|
View Full Product Details