Gene Transcripts
- Gene Symbol
- Myo1b
- Entrez Gene ID
- 117057
- Gene Name
- myosin Ib
- Gene Aliases
- Myr1
- Chromosome Location
- Chr. 9: 54558202 - 54705350 on Build Rnor_6.0
- UniGene ID
- Rn.36339
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053986.1 |
NP_446438.1
|
|
2719 |
|
GenBank mRNA |
X68199.1 |
- |
|
2719 |
|
View Full Product Details