Gene Transcripts

Gene Symbol
Pnma1
Entrez Gene ID
170636
Gene Name
paraneoplastic Ma antigen 1
Gene Aliases
MA1
Chromosome Location
Chr. 6: 107631227 - 107633012 on Build Rnor_6.0
UniGene ID
Rn.81186
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130820.2 NP_570833.1 1 672
GenBank mRNA AF335505.1 - 1 688
BC097291.1 - 1 648 7453131

View Full Product Details