Gene Transcripts

Gene Symbol
Fgf6
Entrez Gene ID
170700
Gene Name
fibroblast growth factor 6
Gene Aliases
Chromosome Location
Chr. 4: 159563798 - 159572333 on Build Rnor_6.0
UniGene ID
Rn.81222
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_131908.1 NP_571983.1 3 470
GenBank mRNA AB079674.1 - 3 470

View Full Product Details