Gene Transcripts

Gene Symbol
Stk17b
Entrez Gene ID
170904
Gene Name
serine/threonine kinase 17b
Gene Aliases
Drak2
Chromosome Location
Chr. 9: 60399261 - 60430081 on Build Rnor_6.0
UniGene ID
Rn.1300
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133392.1 NP_596883.1 4 608
XM_006244908.2 XP_006244970.1 4 1377
GenBank mRNA AB070349.1 - 4 608

View Full Product Details