Gene Transcripts

Gene Symbol
Atg3
Entrez Gene ID
171415
Gene Name
autophagy related 3
Gene Aliases
Apg3l, PIG-1, Pig1
Chromosome Location
Chr. 11: 60585192 - 60613706 on Build Rnor_6.0
UniGene ID
Rn.3084
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134394.2 NP_599221.1 2 377
GenBank mRNA AF175224.1 - 2 380
BC078743.1 - 2 389 7112937

View Full Product Details