Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138508.4 | NP_612517.2 | 8 | 664 | |
GenBank mRNA | BC081713.1 | - | 8 | 677 | |
M34728.1 | - | 2 | 131 | ||
M57453.1 | - | 8 | 664 | ||
M58287.1 | - | 7 | 550 | ||
M62763.1 | - | 8 | 691 | ||
X60654.1 | - | 8 | 644 |