Gene Transcripts

Gene Symbol
ZBTB32
Entrez Gene ID
27033
Gene Name
zinc finger and BTB domain containing 32
Gene Aliases
FAXF, FAZF, Rog, TZFP, ZNF538
Chromosome Location
Chr. 19: 35704481 - 35717048 on Build GRCh38
UniGene ID
Hs.99430
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014383.2 NP_055198.1 3 773
XM_011526718.2 XP_011525020.1 2 934
XM_017026588.1 XP_016882077.1 1 439
XM_017026589.1 XP_016882078.1 3 1144
XM_017026590.1 XP_016882079.1 1 439
GenBank mRNA AF130255.1 - 2 625
AF165097.1 - 1 415
BC017700.2 - 2 541 4419776
KU178599.1 - 1 415
KU178600.1 - 1 415

View Full Product Details