Gene Transcripts

Gene Symbol
Lmo1
Entrez Gene ID
245979
Gene Name
LIM domain only 1
Gene Aliases
Dat1, Lmo3
Chromosome Location
Chr. 1: 173728183 - 173764288 on Build Rnor_6.0
UniGene ID
Rn.25503
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139112.2 NP_620812.2 2 335
XM_006229968.1 XP_006230030.1 3 637
GenBank mRNA BF420017.1 - 2 335
BF567497.1 - 1 147

View Full Product Details