Gene Transcripts

Gene Symbol
Prpf19
Entrez Gene ID
246216
Gene Name
pre-mRNA processing factor 19
Gene Aliases
Prp19
Chromosome Location
Chr. 1: 226947069 - 226958147 on Build Rnor_6.0
UniGene ID
Rn.19573
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139333.3 NP_647549.2 5 606
XM_006231013.2 XP_006231075.1 5 699
XM_006231014.2 XP_006231076.1 5 697
XM_006231015.2 XP_006231077.1 5 641
GenBank mRNA AB020022.2 - 5 652
BC107669.1 - 5 606 7937645

View Full Product Details