Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164073.1 | NP_001157545.1 | 3 | 310 | |
NM_145788.2 | NP_665731.2 | 3 | 321 | ||
XM_006234273.2 | XP_006234335.1 | 3 | 383 | ||
XM_006234274.2 | XP_006234336.1 | 3 | 369 | ||
GenBank mRNA | AJ422211.1 | - | 2 | 181 | |
BC088854.1 | - | 3 | 258 | ||
BC129086.1 | - | 3 | 310 | 7365827 | |
EV776506.1 | - | 3 | 321 |