Gene Transcripts

Gene Symbol
Rab31
Entrez Gene ID
246324
Gene Name
RAB31, member RAS oncogene family
Gene Aliases
Rab0
Chromosome Location
Chr. 9: 113370072 - 113504606 on Build Rnor_6.0
UniGene ID
Rn.28921
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145094.2 NP_659562.2 6 503
GenBank mRNA AF254800.1 - 6 452
BC072698.1 - 6 503 7105732

View Full Product Details