Gene Transcripts

Gene Symbol
Msi1
Entrez Gene ID
259272
Gene Name
musashi RNA-binding protein 1
Gene Aliases
Msi1h
Chromosome Location
Chr. 12: 46899835 - 46935335 on Build Rnor_6.0
UniGene ID
Rn.162146
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148890.1 NP_683688.1 6 488
GenBank mRNA AY043393.1 - 6 488

View Full Product Details