Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153467.1 | NP_703197.1 | 3 | 339 | |
XM_006236545.2 | XP_006236607.1 | 3 | 848 | ||
XM_008762878.1 | XP_008761100.1 | 4 | 775 | ||
GenBank mRNA | AF294688.1 | - | 3 | 339 | |
FQ213688.1 | - | 3 | 301 |