Gene Transcripts

Gene Symbol
Hs3st2
Entrez Gene ID
293451
Gene Name
heparan sulfate (glucosamine) 3-O-sulfotransferase 2
Gene Aliases
Chromosome Location
Chr. 1: 191344979 - 191474143 on Build Rnor_6.0
UniGene ID
Rn.49010
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181370.2 NP_852035.1 2 1375
GenBank mRNA AY240873.1 - 2 1375
BC128721.1 - 2 1378 8364509

View Full Product Details