Gene Transcripts
- Gene Symbol
- Tmsb10
- Entrez Gene ID
- 50665
- Gene Name
- thymosin, beta 10
- Gene Aliases
- Ptmb10, THYb10
- Chromosome Location
- Chr. 4: 100882216 - 100883294 on Build Rnor_6.0
- UniGene ID
- Rn.5983
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021261.2 |
NP_067084.1
|
2, 3 |
180 |
|
XM_006236692.2 |
XP_006236754.1
|
2, 3 |
231 |
|
GenBank mRNA |
BC126092.1 |
- |
2, 3 |
149 |
|
FQ221466.1 |
- |
2, 3 |
178 |
|
FQ223015.1 |
- |
2, 3 |
178 |
|
FQ224120.1 |
- |
2, 3 |
180 |
|
FQ224544.1 |
- |
2, 3 |
178 |
|
FQ229044.1 |
- |
2, 3 |
179 |
|
M17698.1 |
- |
2, 3 |
159 |
|
M58404.1 |
- |
1, 2 |
254 |
|
M58405.1 |
- |
2, 3 |
161 |
|
View Full Product Details