Gene Transcripts

Gene Symbol
Dio1
Entrez Gene ID
25430
Gene Name
deiodinase, iodothyronine, type I
Gene Aliases
5DI, ITDI1, TXDI1
Chromosome Location
Chr. 5: 126895286 - 126911520 on Build Rnor_6.0
UniGene ID
Rn.87549
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021653.3 NP_067685.5 1 215
GenBank mRNA BC083557.1 - 1 210
CK480517.1 - 1 215
X57999.1 - 1 214

View Full Product Details