Gene Transcripts

Gene Symbol
Thoc6
Entrez Gene ID
79227
Gene Name
THO complex 6 homolog (Drosophila)
Gene Aliases
Pdrp
Chromosome Location
Chr. 10: 12989135 - 12994495 on Build Rnor_6.0
UniGene ID
Rn.42184
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024384.2 NP_077360.2 11 1022
GenBank mRNA AF275266.1 - 4 287
BC079149.1 - 11 1022 7128541

View Full Product Details