Gene Transcripts

Gene Symbol
Pvalb
Entrez Gene ID
25269
Gene Name
parvalbumin
Gene Aliases
PALB1, Pva
Chromosome Location
Chr. 7: 119428652 - 119443656 on Build Rnor_6.0
UniGene ID
Rn.2005
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022499.2 NP_071944.1 2 155
XM_006241929.2 XP_006241991.1 3 228
GenBank mRNA BC126090.1 - 2 155
FQ212120.1 - 3 218
FQ214597.1 - 3 216
FQ214667.1 - 3 217
FQ214909.1 - 3 216
FQ215072.1 - 3 216
FQ215101.1 - 1 52
FQ215172.1 - 3 216
FQ215177.1 - 3 215
FQ215250.1 - 3 217
FQ215367.1 - 3 216
FQ215388.1 - 3 216
FQ215455.1 - 3 216
FQ215472.1 - 3 216
FQ215487.1 - 3 216
FQ215608.1 - 3 217
FQ215681.1 - 3 216
FQ216147.1 - 3 216
FQ216186.1 - 3 216
FQ216201.1 - 3 217
FQ216260.1 - 3 216
FQ216338.1 - 3 217
FQ216339.1 - 3 216
FQ216448.1 - 3 216
FQ216459.1 - 3 216
FQ216533.1 - 3 216
FQ216659.1 - 3 216
FQ216789.1 - 3 216
FQ216830.1 - 3 216
FQ216843.1 - 3 216
FQ216867.1 - 3 216
FQ216872.1 - 3 216
FQ216903.1 - 3 218
FQ216906.1 - 3 213
FQ216960.1 - 3 216
FQ216981.1 - 3 216
FQ217053.1 - 3 216
FQ217064.1 - 3 216
FQ217099.1 - 3 216
FQ217132.1 - 2 217
FQ217209.1 - 3 215
FQ217210.1 - 3 216
FQ217278.1 - 3 216
FQ217298.1 - 3 217
FQ217373.1 - 3 217
FQ217384.1 - 3 216
FQ217418.1 - 3 217
FQ217424.1 - 3 216
FQ217446.1 - 2 184
FQ217489.1 - 3 216
FQ217527.1 - 3 216
FQ217562.1 - 3 216
FQ217576.1 - 3 216
FQ217645.1 - 3 215
FQ217652.1 - 3 217
FQ217677.1 - 3 217
FQ217743.1 - 3 215
FQ217749.1 - 3 217
FQ217760.1 - 3 217
FQ217770.1 - 3 216
FQ217787.1 - 3 217
FQ217845.1 - 3 216
FQ217858.1 - 3 216
FQ217860.1 - 3 217
FQ217861.1 - 3 216
FQ217880.1 - 3 218
FQ217886.1 - 3 216
FQ217934.1 - 2 158
FQ217966.1 - 3 216
FQ217976.1 - 3 216
FQ217991.1 - 3 216
FQ218022.1 - 3 216
FQ218035.1 - 3 216
FQ218057.1 - 3 216
FQ223582.1 - 3 216
FQ223596.1 - 3 216
FQ223603.1 - 3 217
FQ223630.1 - 3 213
FQ223664.1 - 3 217
FQ223669.1 - 1 50
FQ223695.1 - 3 216
FQ223712.1 - 3 217
FQ223861.1 - 3 216
FQ223863.1 - 3 216
FQ223941.1 - 3 216
FQ223961.1 - 3 216
FQ223988.1 - 3 215
FQ223989.1 - 3 216
FQ223990.1 - 3 216
FQ224014.1 - 3 216
FQ224025.1 - 3 217
FQ224028.1 - 3 216
FQ224037.1 - 3 216
FQ224053.1 - 3 216
FQ224059.1 - 3 216
FQ224083.1 - 3 216
FQ224113.1 - 3 216
FQ224136.1 - 3 216
FQ224173.1 - 3 216
FQ224181.1 - 3 216
FQ224242.1 - 3 217
FQ224248.1 - 3 214
FQ224294.1 - 3 216
FQ224303.1 - 3 217
FQ224322.1 - 3 216
FQ224356.1 - 3 216
FQ224363.1 - 3 216
FQ224417.1 - 3 216
FQ224430.1 - 3 216
FQ224435.1 - 3 214
FQ224463.1 - 3 217
FQ224483.1 - 3 216
FQ224485.1 - 3 216
FQ224488.1 - 3 217
FQ224510.1 - 3 216
FQ224541.1 - 3 216
FQ224580.1 - 3 216
FQ224584.1 - 3 217
FQ224600.1 - 3 216
FQ224613.1 - 3 216
FQ224634.1 - 3 216
FQ224643.1 - 3 216
FQ224673.1 - 3 217
FQ224675.1 - 3 216
FQ224723.1 - 3 218
FQ224767.1 - 3 216
FQ224778.1 - 3 216
FQ224913.1 - 3 216
M10764.1 - 1 41
M12725.1 - 3 215

View Full Product Details