Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007423.4 | NP_031449.3 | 8 | 1036 | |
GenBank mRNA | AK010934.1 | - | 8 | 1012 | |
AK075972.1 | - | 8 | 1016 | ||
AK076053.1 | - | 8 | 1012 | ||
AK076197.1 | - | 8 | 1014 | ||
AK145975.1 | - | 8 | 1012 | ||
AK146513.1 | - | 8 | 1014 | ||
AK146524.1 | - | 8 | 1014 | ||
AK167549.1 | - | 8 | 1012 | ||
AK168426.1 | - | 8 | 1015 | ||
AK168574.1 | - | 8 | 1014 | ||
AK168576.1 | - | 8 | 1012 | ||
AK168751.1 | - | 8 | 1014 | ||
AK168818.1 | - | 7 | 890 | ||
AK168883.1 | - | 8 | 1012 | ||
AK169046.1 | - | 8 | 1014 | ||
AK169054.1 | - | 8 | 1014 | ||
AK169147.1 | - | 8 | 1012 | ||
BC066206.1 | - | 8 | 999 | 30469872 | |
V00743.1 | - | 8 | 1010 |