Gene Transcripts

Gene Symbol
Bach2
Entrez Gene ID
12014
Gene Name
BTB and CNC homology 2
Gene Aliases
E030004N02Rik
Chromosome Location
Chr. 4: 32238574 - 32586108 on Build GRCm38
UniGene ID
Mm.491223
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109661.1 NP_001103131.1 3 580
GenBank mRNA BC131642.1 - 3 488
CX200493.1 - 1 133

View Full Product Details