Gene Transcripts

Gene Symbol
Bmpr2
Entrez Gene ID
12168
Gene Name
bone morphogenetic protein receptor, type II (serine/threonine kinase)
Gene Aliases
2610024H22Rik, AL117858, AW546137, BB189135, BMP-2, BMPR-2, BMPR-II, BMPRII, BRK-3, Gm20272
Chromosome Location
Chr. 1: 59764270 - 59878081 on Build GRCm38
UniGene ID
Mm.391654
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007561.4 NP_031587.1 13 3846
GenBank mRNA AK148180.1 - 8 2664
U78048.1 - 13 3488

View Full Product Details