Gene Transcripts

Gene Symbol
Cd53
Entrez Gene ID
12508
Gene Name
CD53 antigen
Gene Aliases
AI323659, Ox-44, Tspan25
Chromosome Location
Chr. 3: 106758861 - 106790149 on Build GRCm38
UniGene ID
Mm.316861
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007651.3 NP_031677.1 5 555
GenBank mRNA AA177409.1 - 5 443
AK156510.1 - 5 462
AK160326.1 - 5 453
BC021310.1 - 5 469 5010267
BC052905.1 - 5 420 30078163
X97227.1 - 4 356
Z16071.1 - 4 341

View Full Product Details