Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001285412.1 | NP_001272341.1 | 4 | 483 | |
NM_007594.4 | NP_031620.1 | 4 | 666 | ||
NM_184053.3 | NP_908942.1 | 4 | 666 | ||
GenBank mRNA | AK135295.1 | - | 4 | 591 | |
AK147756.1 | - | 4 | 619 | ||
AK148178.1 | - | 4 | 619 | ||
AK160795.1 | - | 4 | 619 | ||
AK161807.1 | - | 4 | 623 | ||
AK163674.1 | - | 4 | 654 | ||
AK166705.1 | - | 4 | 619 | ||
AY225334.1 | - | 4 | 592 | ||
BC051423.1 | - | 4 | 627 | 3989906 | |
U81829.1 | - | 4 | 592 |