Gene Transcripts

Gene Symbol
Clk4
Entrez Gene ID
12750
Gene Name
CDC like kinase 4
Gene Aliases
AI987988, C85119
Chromosome Location
Chr. 11: 51263167 - 51281770 on Build GRCm38
UniGene ID
Mm.239354
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF033566.1 - 3 333
AK013974.2 - 2 179
AK051099.1 - 1 2439
AY653308.1 - 3 314
BC012675.1 - 3 355 3982920
CT010293.1 - 2 255
U94846.1 - 3 352

View Full Product Details