Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007842.2 | NP_031868.2 | 2, 3 | 269 | |
GenBank mRNA | AK013423.1 | - | 2, 3 | 270 | |
AK141824.1 | - | 2, 3 | 261 | ||
AK148423.1 | - | 2, 3 | 261 | ||
AK169735.1 | - | 2, 3 | 269 | ||
BC033911.1 | - | 2, 3 | 269 | ||
BC089159.1 | - | 2, 3 | 263 | ||
U91922.1 | - | 2, 3 | 226 |