Gene Transcripts

Gene Symbol
GPR61
Entrez Gene ID
83873
Gene Name
G protein-coupled receptor 61
Gene Aliases
BALGR, GPCR3
Chromosome Location
Chr. 1: 109537539 - 109547260 on Build GRCh38
UniGene ID
Hs.709782
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031936.4 NP_114142.3 2 2101
XM_017002438.1 XP_016857927.1 2 4434
GenBank mRNA AF258342.1 - 3 1737
AK290027.1 - 2 2101

View Full Product Details