Gene Transcripts

Gene Symbol
Gsx1
Entrez Gene ID
14842
Gene Name
GS homeobox 1
Gene Aliases
Gsh-1, Gsh1
Chromosome Location
Chr. 5: 147188696 - 147190947 on Build GRCm38
UniGene ID
Mm.4797
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008178.2 NP_032204.1 2 958
GenBank mRNA AB221618.1 - 2 768
BC137769.1 - 2 927 8860788
U21224.1 - 2 958

View Full Product Details