Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021961.5 | NP_068780.2 | 4, 5 | 882 | |
GenBank mRNA | AK310928.1 | - | 2, 3 | 276 | |
AL035973.1 | - | 2, 3 | 167 | ||
AL833289.1 | - | 3, 4 | 416 | ||
BC115398.1 | - | 3, 4 | 375 | 40068609 |