Gene Transcripts

Gene Symbol
Il12rb1
Entrez Gene ID
16161
Gene Name
interleukin 12 receptor, beta 1
Gene Aliases
CD212, IL-12R[b]
Chromosome Location
Chr. 8: 70808360 - 70821424 on Build GRCm38
UniGene ID
Mm.731
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008353.2 NP_032379.2 3 257
GenBank mRNA AK137577.1 - 3 258
AK171937.1 - 3 234
BC057212.1 - 3 240 5350447
U23922.1 - 3 212

View Full Product Details