Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008564.2 | NP_032590.2 | 2 | 339 | |
GenBank mRNA | AK037751.1 | - | 2 | 263 | |
AK077801.1 | - | 2 | 241 | ||
AK088156.1 | - | 2 | 252 | ||
AK088378.1 | - | 2 | 239 | ||
AK129039.1 | - | 2 | 220 | ||
AK145176.1 | - | 2 | 250 | ||
AK146187.1 | - | 2 | 254 | ||
AK146375.1 | - | 2 | 263 | ||
AK180955.1 | - | 1 | 110 | ||
AK184421.1 | - | 1 | 110 | ||
AK187654.1 | - | 1 | 110 | ||
AK190652.1 | - | 1 | 109 | ||
BC055318.1 | - | 2 | 248 | 5696348 | |
BY324067.1 | - | 2 | 339 | ||
D86725.1 | - | 2 | 259 |