Gene Transcripts

Gene Symbol
TEAD1
Entrez Gene ID
7003
Gene Name
TEA domain transcription factor 1
Gene Aliases
AA, NTEF-1, REF1, TCF-13, TCF13, TEAD-1, TEF-1
Chromosome Location
Chr. 11: 12674422 - 12944737 on Build GRCh38
UniGene ID
Hs.655331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021961.5 NP_068780.2 3 795
GenBank mRNA AL035973.1 - 1 80
AL833289.1 - 2 329
BC115398.1 - 2 288 40068609
DR000961.1 - 3 633

View Full Product Details