Gene Transcripts
- Gene Symbol
- Nsg2
- Entrez Gene ID
- 18197
- Gene Name
- neuron specific gene family member 2
- Gene Aliases
- Chromosome Location
- Chr. 11: 32000386 - 32059211 on Build GRCm38
- UniGene ID
- Mm.3304
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290680.1 |
NP_001277609.1
|
2, 3 |
200 |
|
NM_001290681.1 |
NP_001277610.1
|
2, 3 |
196 |
|
NM_008741.4 |
NP_032767.1
|
2, 3 |
336 |
|
GenBank mRNA |
AK011563.1 |
- |
1, 2 |
152 |
|
AK041338.1 |
- |
2, 3 |
254 |
|
AK049679.1 |
- |
2, 3 |
293 |
|
AK075612.1 |
- |
2, 3 |
293 |
|
AK088499.1 |
- |
2, 3 |
328 |
|
BC018224.1 |
- |
2, 3 |
268 |
4527900 |
BU058771.1 |
- |
2, 3 |
199 |
|
DN176718.1 |
- |
2, 3 |
336 |
|
U17259.1 |
- |
2, 3 |
296 |
|
View Full Product Details