Gene Transcripts

Gene Symbol
Pdcd2
Entrez Gene ID
18567
Gene Name
programmed cell death 2
Gene Aliases
AI528543, Pcd2, RP-8
Chromosome Location
Chr. 17: 15521575 - 15527301 on Build GRCm38
UniGene ID
Mm.323
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008799.2 NP_032825.2 2 341
GenBank mRNA AK045985.1 - 2 342
DQ906042.1 - 321
U10903.1 - 2 321

View Full Product Details