Gene Transcripts

Gene Symbol
Pmp22
Entrez Gene ID
18858
Gene Name
peripheral myelin protein 22
Gene Aliases
22kDa, Gas-3, HNPP, Tr, trembler
Chromosome Location
Chr. 11: 63128982 - 63159547 on Build GRCm38
UniGene ID
Mm.1237
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK076005.1 - 4 421
AK171745.1 - 4 418
BC010765.1 - 4 444 4225316
M32240.1 - 4 471
Z38110.1 - 3 285

View Full Product Details