Gene Transcripts

Gene Symbol
Peg3
Entrez Gene ID
18616
Gene Name
paternally expressed 3
Gene Aliases
AL022617, ASF-1, End4, Gcap4, Pw1, Zfp102, mKIAA0287
Chromosome Location
Chr. 7: 6705960 - 6730419 on Build GRCm38
UniGene ID
Mm.389800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008817.2 NP_032843.2 9 1914
GenBank mRNA AB003040.1 - 9 1914
AF038939.1 - 6 1565
AK044602.1 - 10 1978
AK122238.1 - 9 1893
AK141217.1 - 9 1915
AK163845.1 - 9 1917
BC048778.1 - 4 1291 6308481
BC072661.1 - 9 1902 30543809
BC085183.1 - 9 1899 30663888
BC089344.1 - 1 994

View Full Product Details