Gene Transcripts

Gene Symbol
Rps29
Entrez Gene ID
20090
Gene Name
ribosomal protein S29
Gene Aliases
Chromosome Location
Chr. 12: 69157722 - 69159186 on Build GRCm38
UniGene ID
Mm.379006
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009093.2 NP_033119.1 2, 3 298
GenBank mRNA AA217935.1 - 2, 3 182
AK002939.1 - 2, 3 202
AK012285.1 - 2, 3 201
AK224925.1 - 2, 3 191
BC024393.1 - 218 4986748
BC051203.1 - 2, 3 185 1476903
BC055750.1 - 2, 3 249
L31609.1 - 2, 3 182

View Full Product Details